
(S)-4-(tert-Butoxycarbonyl)morpholine-3-carboxylic acid
Catalog # APOSOR308160-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:231.2457 g/mol
Melting Pt:169…173 °C
:GHS07
Storage Temperature:Ambient
CAS Number:783350-37-8
Formula:C₁₀H₁₇NO₅
MDL Number:MFCD04114904