
2,7-Diazaspiro[3.5]nonane-2-carboxylic acid tert-butyl ester 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:236406-55-6
:
Formula:C₁₂H₂₂N₂O₂
MDL Number:MFCD09834803
:226.32 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together