Order Entry
Czech Republic
ContactUsLinkComponent
2,7-Diazaspiro[3.5]nonane-7-carboxylic acid tert-butyl ester 98%
Irritant
2,7-Diazaspiro[3.5]nonane-7-carboxylic acid tert-butyl ester 98%
Catalog # APOSOR308112-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,7-Diazaspiro[3.5]nonane-7-carboxylic acid tert-butyl ester 98%
Catalog # APOSOR308112-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :226.32 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:896464-16-7
Formula:C₁₂H₂₂N₂O₂
MDL Number:MFCD09839640