Order Entry
Czech Republic
ContactUsLinkComponent
tert-Butyl-6-oxo-2-azaspiro[3.3]heptane-2-carboxylate
Irritant
tert-Butyl-6-oxo-2-azaspiro[3.3]heptane-2-carboxylate
Catalog # APOSOR308105-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
tert-Butyl-6-oxo-2-azaspiro[3.3]heptane-2-carboxylate
Catalog # APOSOR308105-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :211.2576 g/mol
Boiling Pt:314.7 °C
Flash Pt:114.1 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1181816-12-5
Formula:C₁₁H₁₇NO₃
MDL Number:MFCD15071430