1H-Pyrrolo[2,3-b]pyridine-5-carbaldehyde
Catalog # APOSOR308040-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:146.15 g/mol
Storage Temperature:Ambient
CAS Number:849067-90-9
Formula:C₈H₆N₂O
MDL Number:MFCD08272241