Order Entry
Czech Republic
ContactUsLinkComponent
2-Chloro-3-(chloromethyl)-5,7-dimethylquinoline
2-Chloro-3-(chloromethyl)-5,7-dimethylquinoline
Catalog # APOSOR307904-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Chloro-3-(chloromethyl)-5,7-dimethylquinoline
Catalog # APOSOR307904-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Storage Temperature:Ambient
CAS Number:948290-59-3
Formula:C₁₂H₁₁Cl₂N