Order Entry
Czech Republic
ContactUsLinkComponent
(-)-(5aS,10bR)-5a,10b-Dihydro-2-(2,4,6-trimethylphenyl)-4H,6H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazinium chloride monohydrate
(-)-(5aS,10bR)-5a,10b-Dihydro-2-(2,4,6-trimethylphenyl)-4H,6H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazinium chloride monohydrate
Catalog # APOSOR307708-250MG
Supplier:  Apollo Scientific
CAS Number:  
(-)-(5aS,10bR)-5a,10b-Dihydro-2-(2,4,6-trimethylphenyl)-4H,6H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazinium chloride monohydrate
Catalog # APOSOR307708-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :367.88 g/mol
Storage Temperature:Ambient
CAS Number:919102-70-8
Formula:C₂₁H₂₂ClN₃O
MDL Number:MFCD10566990