(1R,2S)-Fmoc-2-aminocyclohex-3-enecarboxylic acid
Catalog # APOSOR307664-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:363.42 g/mol
Storage Temperature:Ambient
CAS Number:233600-20-9
Formula:C₂₂H₂₁NO₄
MDL Number:MFCD09750521