1-[Amino(4-chlorophenyl)methyl]-2-naphthol hydrochloride
Catalog # APOSOR307613-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Typ balení:Bottle
- Bal.:1 g
- Sekvence:3047
Specifications
About this item
:
:320.22 g/mol
Storage Temperature:Ambient
CAS Number:856209-29-5
Formula:C₁₇H₁₅Cl₂NO
MDL Number:MFCD07366595