
3-Bromo-1H-pyrazolo[3,4-c]pyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:76006-13-8
:
Formula:C₆H₄BrN₃
MDL Number:MFCD08234919
:198.0200 g/mol
Melting Pt:208…210 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together