
2-Methyl-2H-indazole-4-carboxylic acid methyl ester
Catalog # APOSOR30722-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:190.2 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:1071428-43-7
Formula:C₁₀H₁₀N₂O₂
MDL Number:MFCD11109405