Order Entry
Czech Republic
ContactUsLinkComponent
 
cis-4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid 98%
Catalog # APOSOR307110-10G
Supplier:  Apollo Scientific
CAS Number:  
cis-4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid 98%
Catalog # APOSOR307110-10G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :243.30 g/mol
Storage Temperature:Ambient
CAS Number:53292-90-3
Formula:C₁₂H₂₁NO₄
MDL Number:MFCD01862294