Order Entry
Czech Republic
ContactUsLinkComponent
(1S,2R)-cis-4-Cyclohexene-1,2-dicarboxylic acid 1-monomethyl ester
Irritant
(1S,2R)-cis-4-Cyclohexene-1,2-dicarboxylic acid 1-monomethyl ester
Catalog # APOSOR307028-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
(1S,2R)-cis-4-Cyclohexene-1,2-dicarboxylic acid 1-monomethyl ester
Catalog # APOSOR307028-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :184.19 g/mol
Melting Pt:65…67 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:88335-93-7
Formula:C₉H₁₂O₄
MDL Number:MFCD00075490