
5-Hexyn-1-ol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:928-90-5
:
Formula:C₆H₁₀O
MDL Number:MFCD00002980
:98.14 g/mol
Boiling Pt:73…75 °C/15mmHg
Density:0.89 g/cm³ (25 °C)
Flash Pt:70 °C
EINECS:618-884-7
:GHS07
Storage Temperature:Ambient
Frequently Bought Together