

4,6-Dichloro-o-cresol
:1570-65-6
:
:C₇H₆Cl₂O
:MFCD00019988
:177.03 g/mol
:225…226 °C
:53…56 °C
:216-382-9
:GHS07
:GHS05
:Ambient
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Ammonium acetate ≥99%, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals