Order Entry
Czech Republic
ContactUsLinkComponent
 
Irritant
4-Bromo-o-toluonitrile
Catalog # APOSOR3057-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Bromo-o-toluonitrile
Catalog # APOSOR3057-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:63…64 °C
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
 :196.05 g/mol
 :GHS07
CAS Number:67832-11-5
Formula:C₈H₆BrN
MDL Number:MFCD00041461