1,3-Dimethyl-1H-pyrazole-4-carboxylic acid
Catalog # APOSOR305621-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:140.14 g/mol
Storage Temperature:Ambient
CAS Number:78703-53-4
Formula:C₆H₈N₂O₂
MDL Number:MFCD00464005