cis-N-Boc-4-cyano-L-proline methyl ester
Catalog # APOSOR305322-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:254.286 g/mol
Storage Temperature:Ambient
CAS Number:487048-28-2
Formula:C₁₂H₁₈N₂O₄
MDL Number:MFCD03787928