
(2-Methoxyphenyl)hydrazine hydrochloride 96%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:6971-45-5
:
Formula:C₇H₁₁ClN₂O
MDL Number:MFCD00035456
:174.6280 g/mol
Melting Pt:155 °C
EINECS:230-199-1
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together