
(1R,2S)-1-[(tert-Butoxycarbonyl)amino]-2-vinylcyclopropanecarboxylic acid
Catalog # APOSOR305053-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:227.26 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:159622-10-3
Formula:C₁₁H₁₇NO₄
MDL Number:MFCD11042412