
Ethyl-2-amino-4-chlorothieno[2,3-d]-pyrimidine-6-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:847560-46-7
:
Formula:C₉H₈ClN₃O₂S
MDL Number:MFCD18074287
:257.7 g/mol
Melting Pt:253…255 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together