
(6-Morpholino-3-pyridinyl)-1-ethanone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:265107-43-5
:
Formula:C₁₁H₁₄N₂O₂
MDL Number:MFCD09910348
Melting Pt:78…80 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together