6-[(4-Methoxyphenyl)sulphanyl]-3-pyridinylamine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1019363-45-1
:
Formula:C₁₂H₁₂N₂OS
MDL Number:MFCD11132352
:232.31 g/mol
Melting Pt:67…69 °C
Storage Temperature:Ambient
Frequently Bought Together