
(3,4-Dinitrophenyl)methanol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:79544-31-3
:
Formula:C₇H₆N₂O₅
MDL Number:MFCD00007094
:198.14 g/mol
Melting Pt:57…59 °C
EINECS:279-178-9
:GHS07
Storage Temperature:Ambient
Frequently Bought Together