

1-Iodo-2-naphthol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2033-42-3
:
Formula:C₁₀H₇IO
MDL Number:MFCD00046371
:270.07 g/mol
Boiling Pt:307 °C/760 mmHg
Melting Pt:92…96 °C
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together