
1-Methyl-3-thien-2-yl-1H-pyrazol-5-amine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:118430-78-7
:
Formula:C₈H₉N₃S
MDL Number:MFCD00068075
:179.25 g/mol
Melting Pt:137…140 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together