2-[(4-Methylphenyl)sulphanyl]-5-nitrobenzenecarbaldehyde
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:175278-42-9
:
Formula:C₁₄H₁₁NO₃S
MDL Number:MFCD00173708
:273.31 g/mol
Melting Pt:155…157 °C
Storage Temperature:Ambient
Frequently Bought Together