Order Entry
Czech Republic
ContactUsLinkComponent
tert-Butyl-4-{[8-(ethoxycarbonyl)[1,2,4]triazolo[4,3-a]pyridin-3-yl]methyl}piperidine-1-carboxylate
tert-Butyl-4-{[8-(ethoxycarbonyl)[1,2,4]triazolo[4,3-a]pyridin-3-yl]methyl}piperidine-1-carboxylate
Catalog # APOSOR303120-500MG
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl-4-{[8-(ethoxycarbonyl)[1,2,4]triazolo[4,3-a]pyridin-3-yl]methyl}piperidine-1-carboxylate
Catalog # APOSOR303120-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :388.47 g/mol
Storage Temperature:Ambient
CAS Number:1211592-94-7
Formula:C₂₀H₂₈N₄O₄
MDL Number:MFCD12755391