tert-Butyl-3-[methoxy(methyl)carbamoyl]cyclohexane-1-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1393845-80-1
:
Formula:C₁₄H₂₅NO₄
MDL Number:MFCD22383958
:271.36 g/mol
Storage Temperature:Ambient
Frequently Bought Together