3-Methoxypiperidine hydrochloride
Catalog # APOSOR302757-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:151.64 g/mol
Storage Temperature:Ambient
CAS Number:688809-94-1
:MFCD07781094
Formula:C₆H₁₃NO·HCl
MDL Number:MFCD06800959