Order Entry
Czech Republic
ContactUsLinkComponent
(9aS)-Octahydropiperazino[2,1-c]morpholine dihydrochloride 97%
(9aS)-Octahydropiperazino[2,1-c]morpholine dihydrochloride 97%
Catalog # APOSOR302681-1G
Supplier:  Apollo Scientific
CAS Number:  
(9aS)-Octahydropiperazino[2,1-c]morpholine dihydrochloride 97%
Catalog # APOSOR302681-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :215.121 g/mol
Storage Temperature:Ambient
CAS Number:1089280-14-7
Formula:C₇H₁₄N₂O·2HCl
MDL Number:MFCD23105798