Order Entry
Czech Republic
ContactUsLinkComponent
 
Ethyl-2,7-dimethylimidazo[1,2-a]pyridine-3-carboxylate
Catalog # APOSOR302245-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl-2,7-dimethylimidazo[1,2-a]pyridine-3-carboxylate
Catalog # APOSOR302245-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :218.2500 g/mol
Storage Temperature:Ambient
CAS Number:81448-48-8
Formula:C₁₂H₁₄N₂O₂
MDL Number:MFCD00139510