2-Hydroxy-5-nitro-6-picoline
Catalog # APOSOR302172-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:154.13 g/mol
Storage Temperature:Ambient
CAS Number:28489-45-4
Formula:C₆H₆N₂O₃
MDL Number:MFCD00092010