5-Bromo-3-methyl-1H-pyrazolo[3,4-b]pyridine
Catalog # APOSOR302144-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:212.0500 g/mol
Storage Temperature:Ambient
CAS Number:885223-65-4
Formula:C₇H₆BrN₃
MDL Number:MFCD11846319