
(1H-Pyrazol-4-yl)methanol
Catalog # APOSOR302030-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:98.1032 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:25222-43-9
Formula:C₄H₆N₂O
MDL Number:MFCD06797673