Order Entry
Czech Republic
ContactUsLinkComponent
2-Bromobenzenesulphonamide 98%
Irritant
2-Bromobenzenesulphonamide 98%
Catalog # APOSOR30195-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Bromobenzenesulphonamide 98%
Catalog # APOSOR30195-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :236.09 g/mol
Melting Pt:191…192 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:92748-09-9
Formula:C₆H₆BrNO₂S
MDL Number:MFCD00173662