

4-Cyanobenzenesulphonyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:49584-26-1
:
Formula:C₇H₄ClNO₂S
MDL Number:MFCD00052337
:201.63 g/mol
Melting Pt:107…112 °C
UN:3261
ADR:8,III
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together