Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 4-chlorobenzoate
Methyl-4-chlorobenzoate
Methyl-4-chlorobenzoate
Irritant
Methyl-4-chlorobenzoate
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Methyl-4-chlorobenzoate
Supplier:
Apollo Scientific
About this item
CAS Number:
1126-46-1
:
Formula:
C₈H₇ClO₂
MDL Number:
MFCD00000621
:
170.6 g/mol
:
GHS07
Storage Temperature:
Ambient
Frequently Bought Together
2,2,4-Trimethylpentane, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Methanol, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Formaldehyde ≥37% in aqueous solution stabilised, GR ACS, Reag. Ph. Eur. analytical reagent, Sigma-Aldrich®
Supplier: Merck
Dichloromethane stabilised, EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®
Supplier: Merck
n-Hexane, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Diethyl ether stabilised, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.9%, HiPerSolv CHROMANORM®, ULTRA for LC-MS, suitable for UPLC/UHPLC-MS instruments
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
tert-Butyl methyl ether, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
Tetrahydrofuran stabilised, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck