Order Entry
Czech Republic
ContactUsLinkComponent
(3-Chloropyrazin-2-yl)methanamine hydrochloride 96%
Irritant
(3-Chloropyrazin-2-yl)methanamine hydrochloride 96%
Catalog # APOSOR301313-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
(3-Chloropyrazin-2-yl)methanamine hydrochloride 96%
Catalog # APOSOR301313-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :180.0352 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:939412-86-9
Formula:C₅H₇Cl₂N₃
MDL Number:MFCD09910171