
5-Bromo-2-methyl-7-azaindole
Catalog # APOSOR301272-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:211.0600 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1111638-02-8
Formula:C₈H₇BrN₂
MDL Number:MFCD15529124