
4-Chlorofuro[3,2-d]pyrimidine
Catalog # APOSOR301170-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:154.5538 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:655255-09-7
Formula:C₆H₃ClN₂O
MDL Number:MFCD09834944