Order Entry
Czech Republic
ContactUsLinkComponent
 
2,4-Dichloro-5,7-dihydrofuro[3,4-d]pyrimidine
Catalog # APOSOR301154-1G
Supplier:  Apollo Scientific
CAS Number:  
2,4-Dichloro-5,7-dihydrofuro[3,4-d]pyrimidine
Catalog # APOSOR301154-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :191.0200 g/mol
Storage Temperature:Ambient
CAS Number:848398-41-4
Formula:C₆H₄Cl₂N₂O
MDL Number:MFCD13250060