1-(4-(Bromomethyl)phenyl)ethanone
Catalog # APOSOR301151-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:213.0700 g/mol
Storage Temperature:Ambient
CAS Number:51229-51-7
Formula:C₉H₉BrO
MDL Number:MFCD10001463