
5-Methoxy-1H-pyrrolo[3,2-b]pyridine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:17288-40-3
:
Formula:C8H8N2O
MDL Number:MFCD08688601
:148.1619 g/mol
Melting Pt:108…112 °C
Storage Temperature:Refrigerator
UN:1759
ADR:8,III
:GHS05
Frequently Bought Together