
3-Pyrrolidinone hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3760-52-9
:
Formula:C₄H₈ClNO
MDL Number:MFCD02183811
:121.5654 g/mol
Melting Pt:143…144 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together