
3-Methoxyazetidine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:148644-09-1
:
Formula:C4H10ClNO
MDL Number:MFCD06804514
:123.58 g/mol
Melting Pt:143…149 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together