
2-Methylpropane-2-sulphinamide 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:146374-27-8
:
Formula:C₄H₁₁NOS
MDL Number:MFCD01863616
:121.20 g/mol
Melting Pt:97…101 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together