Order Entry
Czech Republic
ContactUsLinkComponent
4-Aminocyclopentene hydrochloride
Irritant
4-Aminocyclopentene hydrochloride
Catalog # APOSOR301049-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Aminocyclopentene hydrochloride
Catalog # APOSOR301049-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :119.6 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:91469-55-5
Formula:C₅H₁₀ClN
MDL Number:MFCD01630740