
5,6,7,8-Tetrahydro[1,2,4]triazolo[4,3-a]pyrazine hydrochloride
Catalog # APOSOR301013-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:160.61 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:837430-14-5
Formula:C₅H₉ClN₄
MDL Number:MFCD08448159