Order Entry
Czech Republic
ContactUsLinkComponent
4-Chloro-2,6-dimethoxypyrimidine
4-Chloro-2,6-dimethoxypyrimidine
Catalog # APOSOR301002-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Chloro-2,6-dimethoxypyrimidine
Catalog # APOSOR301002-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :174.59 g/mol
Storage Temperature:Ambient
CAS Number:6320-15-6
Formula:C₆H₇ClN₂O₂
MDL Number:MFCD00006068